Learn More
Creative Biomart Sargramostim; GM-CSF
Supplier:  Creative Biomart THP0003
Encompass_Preferred
The product is a human recombinant granulocyte macrophage colony-stimulating factor (GM-CSF) expressed in yeast. It is a glycoprotein that is 127 residues. Substitution of Leu23 leads to a difference from native protein.
Catalog No. 50-208-4054