Learn More
STEMCELL Technologies Human Recombinant Heregulin-beta 1, Size: 10 µg
SDP
Supplier:  STEMCELL Technologies 78071
Encompass
Use heregulin-beta 1 in the growth and maintenance of human embryonic stem cells (Wang et al. Blood, 2007). Heregulin-beta 1, also known as neuregulin-1 (NRG-1), is a member of the epidermal growth factor (EGF) family of growth factors and acts as a ligand for ErbB family receptor tyrosine kinases (Britsch et al. Genes Dev, 1998). Heregulin-beta 1 is expressed in neuronal cells and modulates cell growth and differentiation of the cells during development and wound healing (Mei & Xiong. Nat Rev Neurosci 2008). Human recombinant heregulin-beta 1 from STEMCELL comes lyophilized with ≥ 95% purity; EC50 ≤ 0.1 ng/mL; endotoxin levels verified ≤ 0.2 EU/μg protein. Alternative names: Acetylcholine receptor-inducing activity, ARIA, Breast cancer cell differentiation factor p45, Glial growth factor, Heregulin Neu differentiation factor, HRG, HRG1, HRG1-beta 1, Neuregulin-1, NRG1, NRG1-beta 1, Sensory and motor neuron-derived factor. Visit STEMCELL.com for more information.
Catalog No. 50-197-6404
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov