Learn More
STEMCELL Technologies Human Recombinant HGF, Size: 50 µg
SDP
Supplier:  STEMCELL Technologies 78019
Encompass
Hepatocyte growth factor/scatter factor (HGF/SF) is a pleiotropic growth factor that promotes proliferation, motility, survival, and differentiation. HGF binds and promotes the dimerization and activation of the receptor tyrosine kinase c-MET, and stimulates PI3K/AKT, FAK, JNK, and ERK1/2 signaling (Organ & Tsao). HGF stimulates migration of cells during embryogenesis, induces cell motility and scattering of epithelial cells, and regulates epithelial-mesenchymal transition. It is a chemoattractant for motor neurons, and regulates the development of sensory, sympathetic, parasympathetic, and cortical neurons as well as the proliferation of oligodendrocytes and glial development. Additionally, HGF is cytoprotective and regulates liver growth and regeneration (Nakamura et al.).
Catalog No. 50-197-6282
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov