Learn More
STEMCELL Technologies Human Recombinant IFN-alpha 2A, Size: 20 µg
SDP
Supplier:  STEMCELL Technologies 78076
Encompass
Interferon-alpha (IFN-α) is a type I interferon, produced by virus-infected cells, and is released as a soluble factor to initiate antiviral responses (Isaacs & Lindenmann). IFN-α2 is the most potent IFN-α used in fundamental research and in most clinical applications. The best-known IFN-α2 subvariants, 2A and 2B, differ by only one or two amino acids at positions 23 and/or 34 of the mature protein (von Gabain et al.). Type I IFNs exert potent antitumor activity by increasing the cytotoxic activity of NK and T cells, as well as by inhibiting the proliferation of cancer cells (Paul et al.). Additionally, it has been shown that proinflammatory IFN-α modulates the function of B cells in patients with systemic lupus erythematosus (Chang et al.), and pegylated forms of IFN-alpha 2A and 2B have implications in the treatment of hepatitis C (Foster et al.).
Catalog No. 50-197-6414
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov