Learn More
STEMCELL Technologies Human Recombinant IGF-I, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 78022.1
Encompass
Insulin-like growth factor 1 (IGF-I) is a polypeptide that belongs to the family of insulin-like growth factors that are similar in molecular structure to proinsulin. IGF-I binds to the IGF-I receptor and is a potent activator of the PI3K/AKT pathway and also activates ERK1/2 signaling. IGF-I is required for embryonic development, and it is produced mainly in the liver in response to a hepatocyte growth hormone. In the absence of insulin, IGF-I is necessary for the maintenance of human pluripotent stem cells (Wang et al.). Together with IL-3, IGF-I stimulates differentiation and proliferation of myeloid cells and has been shown to regulate lymphopoiesis by stimulating proliferation and differentiation of T and B cells in lymphoid organs (Heemskerk et al.).
Catalog No. 50-197-6288