Learn More
STEMCELL Technologies Human Recombinant IP-10 (CXCL10), Size: 50 µg
SDP
Supplier:  STEMCELL Technologies 78165.1
Encompass
Interferon gamma-inducible protein (IP) 10 or CXCL10 is a member of the CXC chemokine family. It binds CXCR3 activating ERK1/2, p38/MAPK, JNK, and PI3-kinase/AKT signaling pathways, inducing intracellular calcium influx, DNA synthesis, cell proliferation, and chemotaxis. IP-10 regulates innate and adaptive immune responses by affecting the function of activated T cells, natural killer cells, inflammatory dendritic cells, macrophages, and B cells. IP-10 is produced by leukocytes, activated neutrophils, eosinophils, monocytes, epithelial cells, endothelial cells, fibroblasts, and keratinocytes in response to IFN-gamma. IP-10 has been implicated in a wide range of diseases, including infectious diseases and chronic inflammatory and autoimmune diseases, as well as in tumor formation (Liu et al.).
Catalog No. 50-197-6631