Learn More
STEMCELL Technologies Human Recombinant MCP-1 (CCL2), Size: 5 ug
SDP
Supplier:  STEMCELL Technologies 78087
Encompass
Use monocyte chemotactic protein-1 (MCP-1) to activate signal transduction pathways that lead to monocyte transmigration (Sozzani et al. J Immunol 1994). MCP-1 also known as CCL2 is primarily induced by platelet-derived growth factor gene (Cochran et al. Cell 1983). MCP-1 plays a role in neurogenesis neuroprotection and neurotransmission and has implications in different neurological disorders (Conductier et al. J Neuroimmunol 2010). MCP-1 is produced by a variety of cell types including fibroblasts endothelial epithelial monocytic and microglial cells (Deshmane et al. J Interferon Cytokine Res 2009). Human recombinant MCP-1 from STEMCELL comes lyophilized with ≥ 95% purity specific activity EC50 100 ng/mL endotoxin levels verified 1 EU/μg protein. Alternative names DIA Differentiation-inducing factor HILDA Human interleukin in DA cells Leukemia inhibitory factor MCAF Melanoma-derived LPL inhibitor
Catalog No. 50-239-0368