Learn More
STEMCELL Technologies Human Recombinant SCF, ACF, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 78155.1
Encompass
Use stem cell factor (SCF) to promote cell survival, proliferation, differentiation, adhesion, and functional activation of cells at multiple levels of the hematopoietic hierarchy. SCF is an early-acting cytokine that activates PI3K, JAK/STAT, and MAPK pathways upon binding to its receptor. Together with thrombopoietin and Flt3/Flk-2 Ligand, SCF is used to promote expansion of primitive hematopoietic stem cells and multipotent progenitor cells in culture (Martin et al. Cell, 1990; Kent et al. Clin Cancer Res, 2008.). In combination with various growth factors and erythropoietin, SCF increases proliferation and differentiation of myeloid and erythroid progenitor cells and a subset of lymphoid progenitor cells (Broudy VC. Blood, 1997). Human recombinant SCF from STEMCELL is animal component-free and comes lyophilized with ≥ 95% purity; EC50 ≤ 15 ng/mL; endotoxin levels verified ≤ 1 EU/μg protein. Alternative names: Kit ligand, Mast cell growth factor, Steel factor, Stem cell factor.
Catalog No. 50-197-6607
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov