Learn More
STEMCELL Technologies Human Recombinant Shh (C24II), Size: 1000 µg
SDP
Catalog No. 501976390
Encompass
Sonic hedgehog (Shh) is a member of the Hedgehog family of secreted signaling proteins that has a prominent role in patterning during early mammalian development (Ho & Scott). It is expressed during early embryogenesis in a variety of tissues (Finco et al.). During patterning of the central nervous system, Shh directly acts on cells to specify neural cell fates, regulates proliferation and survival of oligodendrocytes precursors and neural crest cells and stimulates proliferation, differentiation and axon growth (Ho & Scott; Martí). Shh also regulates T cell differentiation and activation (Crompton et al.). Recombinant human Shh (C24II; rhShh) is a fully biologically active molecule.
Catalog No. 50-197-6390 Supplier STEMCELL Technologies Supplier No. 78065.2
Sonic hedgehog (Shh) is a member of the Hedgehog family of secreted signaling proteins that has a prominent role in patterning during early mammalian development (Ho & Scott). It is expressed during early embryogenesis in a variety of tissues (Finco et al.). During patterning of the central nervous system, Shh directly acts on cells to specify neural cell fates, regulates proliferation and survival of oligodendrocytes precursors and neural crest cells and stimulates proliferation, differentiation and axon growth (Ho & Scott; Martí). Shh also regulates T cell differentiation and activation (Crompton et al.). Recombinant human Shh (C24II; rhShh) is a fully biologically active molecule.