Learn More
STEMCELL Technologies Human Recombinant TGF-alpha, Size: 10 µg
SDP
Supplier:  STEMCELL Technologies 78123
Encompass
Transforming growth factor-alpha (TGF-α) a member of the epidermal growth factor (EGF) family that is expressed in normal epithelial cells, monocytes, macrophages, brain cells, keratinocytes, and cancer cells. TGF-α binds EGF receptor (EGFR) and activates tyrosine kinase signaling. In epidermal and epithelial cells, this results in proliferation and differentiation. TGF-α is not structurally or genetically related to TGF-β, and the two ligands act through different signaling pathways. In vitro, TGF-α was shown to stimulate anchorage-independent growth (Singh & Coffey).
Catalog No. 50-197-6519