Learn More
Innovative Research Inc Human Red Blood Cell CPDA1 Unit, Single donor, 250 ml volume, CPDA-1 anticoagulant, Centrifugation method, Custom collection
Catalog No. NC3595435
Encompass
This technical summary outlines the key features and attributes of the Human Red Blood Cell CPDA1 Unit, designed specifically for scientific research and manufacturing applications. The product consists of single-donor human red blood cells collected in CPDA-1 anticoagulant, with careful methods employed to ensure quality and compliance with FDA regulations. This custom-collected product allows for flexibility in demographic and collection protocols, ensuring its suitability for a range of experimental purposes.
  • Single human donor source
  • Approximately 250 ml of unwashed and resuspended red blood cells
  • Using CPDA-1 anticoagulant
  • Processed through centrifugation and manual extraction
  • Negative viral testing for multiple pathogens
  • Strictly for in-vitro use in research and manufacturing
Catalog No. NC3595435 Supplier Innovative Research Inc Supplier No. IWB3CPDA1UNINT
This technical summary outlines the key features and attributes of the Human Red Blood Cell CPDA1 Unit, designed specifically for scientific research and manufacturing applications. The product consists of single-donor human red blood cells collected in CPDA-1 anticoagulant, with careful methods employed to ensure quality and compliance with FDA regulations. This custom-collected product allows for flexibility in demographic and collection protocols, ensuring its suitability for a range of experimental purposes.
  • Single human donor source
  • Approximately 250 ml of unwashed and resuspended red blood cells
  • Using CPDA-1 anticoagulant
  • Processed through centrifugation and manual extraction
  • Negative viral testing for multiple pathogens
  • Strictly for in-vitro use in research and manufacturing