Learn More
MOLECULAR DEPOT LLC Human Red Blood Cells - 500 mg
Supplier:  MOLECULAR DEPOT LLC B2013327
Encompass_Preferred
Lyophilized Human Red Blood Cells consists of pooled human red blood cells dextran-separated not washed and lyophilized About 2 5 mL of red blood cells have been used to manufacture this product For in vitro research use only
Catalog No. 50-000-13437