Learn More
Agilent Technologies Stratagene QPCR Human Reference Total RNA
Catalog No. NC9100978
Encompass
Stratagene QPCR Human Reference Total RNA, 25microgram, QPCR Human Reference Total RNA, Concentration: 1microgram/microliter. High-quality total RNA control for quantitative PCR gene-expression analysis. No detectable levels of genomic DNA
Catalog No. NC9100978 Supplier Agilent Technologies Supplier No. 750500
Stratagene QPCR Human Reference Total RNA, 25microgram, QPCR Human Reference Total RNA, Concentration: 1microgram/microliter. High-quality total RNA control for quantitative PCR gene-expression analysis. No detectable levels of genomic DNA