Learn More
Sino Biological Human REG1A MAb 100ul (Sino Biological)
Supplier:  Sino Biological 204456T08100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human REG1A (rh REG1A; Catalog#11234-H08H; NP_002900.2; Met1-Asn166). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-4302