Learn More
Sino Biological Recombinant Human REG1A Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11234H08HB100
Encompass_Preferred
A DNA sequence encoding the human REG1A (NP_002900.2) (Met1-Asn166) precursor was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4759