Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Reg3A ELISA Kit

Catalog No. EH390RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH390RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH390RBX10 Supplier Invitrogen™ Supplier No. EH390RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human Reg3A quantitates human Reg3A in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Reg3A.

REG3A encodes a pancreatic secretory protein that may be involved in cell proliferation or differentiation. It has similarity to the C-type lectin superfamily. The enhanced expression of this protein is observed during pancreatic inflammation and liver carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q06141
Assay Range 0.12-25 ng/mL
Assay Sensitivity 0.12 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AV063448; generating islet-derived, mouse homolog 3 alpha; hepatocarcinoma-intestine-pancreas; hepatointestinal pancreatic protein; HIP; HIP/PAP; human proislet peptide; INGAP; islet of Langerhans regenerating protein 3; Lithostathine 3; pancreatic beta cell growth factor; pancreatitis-associated protein 1; Pancreatitis-associated protein 2; PAP; PAP homologous protein; PAP1; Pap2; PAP-H; PapII; PBCGF; proliferation-inducing protein 34; proliferation-inducing protein 42; REG 3; REG 3a; REG 3alpha; Reg IIIalpha; Reg III-alpha; REG3; REG3 alpha; Reg3a; REG3alpha; REG-3alpha; REG-3-alpha; regeneratin; regenerating family member 3 alpha; regenerating islet-derived 3 alpha; regenerating islet-derived protein 3 alpha; regenerating islet-derived protein 3-alpha; Regenerating islet-derived protein 3-alpha 15 kDa form; Regenerating islet-derived protein 3-alpha 16.5 kDa form; Regenerating islet-derived protein III-alpha; regIII; REG-III; RegIII (alpha)AV063448; generating islet-derived, mouse homolog 3 alpha; hepatocarcinoma-intestine-pancreas; hepatointestinal pancreatic protein; HIP; HIP/PAP; human proislet peptide; INGAP; islet of Langerhans regenerating protein 3; Lithostathine 3; pancreatic beta cell growth factor; pancreatitis-associated protein 1; Pancreatitis-associated protein 2; PAP; PAP homologous protein; PAP1; Pap2; PAP-H; PapII; PBCGF; proliferation-inducing protein 34; proliferation-inducing protein 42; REG 3; REG 3a; REG 3alpha; Reg IIIalpha; Reg III-alpha; REG3; REG3 alpha; Reg3a; REG3alpha; REG-3alpha; REG-3-alpha; regeneratin; regenerating family member 3 alpha; regenerating islet-derived 3 alpha; regenerating islet-derived protein 3 alpha; regenerating islet-derived protein 3-alpha; Regenerating islet-derived protein 3-alpha 15 kDa form; Regenerating islet-derived protein 3-alpha 16.5 kDa form; Regenerating islet-derived protein III-alpha; regIII; REG-III; RegIII (alpha);
Gene ID (Entrez) 5068
Gene Symbol Reg3a
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target REG3A
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.