Learn More
Sino Biological Anti-Reg3A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11235T2620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human REG3A (rh REG3A; Catalog#11235-H08H; Q06141-1; Met1-Asp175). REG3A specific IgG was purified by Human REG3A affinity chromatography.
Catalog No. 50-255-4753