Learn More
Sino Biological Recombinant Human REG3G Protein (His Tag)
Supplier:  Sino Biological 11641H08H50
Encompass_Preferred
A DNA sequence encoding the human REG3G (NP_001008388.1) (Met 1-Asp 175) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7205