Learn More
Sino Biological Anti-REG4 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11186MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human REG4 (rh REG4; Catalog#11186-H08H; NP_001152824.1; Met1-Pro158). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8479