Learn More
Sino Biological Anti-REG4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11186RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human REG4 (rh REG4; Catalog#11186-H08H; NP_001152824.1; Met 1-Pro 158). REG4 specific IgG was purified by human REG4 affinity chromatography.
Catalog No. 50-240-8482