Learn More
Biomatik Corporation Human Regenerating Islet Derived Protein 3 Gamma (REG3g) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU11536
Encompass_Preferred
Human Regenerating Islet Derived Protein 3 Gamma (REG3g) Wide-range ELISA Kit for detecting REG3g in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6UW15. Gene ID: 130120. Target Synonyms: REG-III; PAP1B; PAPIB; UNQ429; Pancreatitis-associated protein 1B; Regenerating islet-derived protein III-gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5781