Learn More
Biomatik Corporation Human Regulated On Activation In Normal T-Cell Expressed And Secreted (RANTES) CLIA Kit
Supplier:  Biomatik Corporation EKU08833
Encompass_Preferred
Human Regulated On Activation In Normal T-Cell Expressed And Secreted (RANTES) CLIA Kit for detecting RANTES in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 11.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P13501. Gene ID: 6352. Target Synonyms: CCL5; SCYA5; SISd; EoCP; Chemokine C-C-Motif Ligand 5; Eosinophil Chemotactic Cytokine; T-cell Specific Protein p288; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8616