Learn More
Biomatik Corporation Human Regulator Of G Protein Signaling 7 (RGS7) ELISA Kit
Supplier:  Biomatik Corporation EKU11394
Encompass_Preferred
Human Regulator Of G Protein Signaling 7 (RGS7) ELISA Kit for detecting RGS7 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49802. Gene ID: 6000. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8149