Learn More
Biomatik Corporation Human Regulator Of G Protein Signaling 19 (RGS19) ELISA Kit
Supplier:  Biomatik Corporation EKU08305
Encompass_Preferred
Human Regulator Of G Protein Signaling 19 (RGS19) ELISA Kit for detecting RGS19 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.12-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49795. Gene ID: 10287. Target Synonyms: GAIP; RGSGAIP; GNAI3IP; G-alpha-interacting protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8681