Learn More
Sino Biological Anti-RELA/NF-kappa B p65 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12054MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human RELA / Transcription factor p65 (rh RELA / Transcription factor p65; Catalog#12054-H09E; Q04206-1; Met 1-Tyr 306). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8720