Learn More
Sino Biological Phospho-NF-κB p65 (Ser536) Antibody, Rabbit MAb
Catalog No. 502993540
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic phosphopeptide corresponding to residues around Ser536 of the Human NF-κB p65.
Catalog No. 50-299-3540 Supplier Sino Biological Supplier No. 111378R000220
This antibody was obtained from a rabbit immunized with a synthetic phosphopeptide corresponding to residues around Ser536 of the Human NF-κB p65.