Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Relaxin-1 ELISA Kit

Catalog No. EH391RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH391RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH391RBX10 Supplier Invitrogen™ Supplier No. EH391RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human Relaxin-1 quantitates human Relaxin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Relaxin-1.

Relaxins are known endocrine and autocrine/paracrine hormones, belonging to the insulin gene superfamily. In the human there are three non-allelic relaxin genes, RLN1, RLN2 and RLN3. RLN1 and RLN2 share high sequence homology. This encoded protein is synthesized as a single-chain polypeptide but the active form consists of an A chain and a B chain linked by disulfide bonds; however, their exact cleavage sites have not been described. Relaxin is produced by the ovary, and targets the mammalian reproductive system to ripen the cervix, elongate the pubic symphysis and inhibit uterine contraction. It may have additional roles in enhancing sperm motility, regulating blood pressure, controlling heart rate and releasing oxytocin and vasopressin. This gene has multiple polyadenylation sites. There are multiple alternatively spliced transcript variants described for this gene but their full length nature is not known yet.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04808
Assay Range 0.23-50 ng/mL
Assay Sensitivity 0.23 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias bA12D24.3.1; bA12D24.3.2; H1; H1RLX; preprorelaxin; preprorelaxin H1; prorelaxin 1; Prorelaxin H1; RELAX; relaxin 1; Relaxin 1 (H1); Relaxin A chain; Relaxin B chain; relaxin RLXH1; Rln; Rln1; rlx; RLX bA12D24.3.1; RLXH1bA12D24.3.1; bA12D24.3.2; H1; H1RLX; preprorelaxin; preprorelaxin H1; prorelaxin 1; Prorelaxin H1; RELAX; relaxin 1; Relaxin 1 (H1); Relaxin A chain; Relaxin B chain; relaxin RLXH1; Rln; Rln1; rlx; RLX bA12D24.3.1; RLXH1;
Gene ID (Entrez) 6013
Gene Symbol Rln1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Relaxin 1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.