Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Relaxin-3 Recombinant Protein, PeproTech®

Catalog No. 130105UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
130-10-5UG 5 μg
13010100UG 100 μg
130-10-1MG 1 mg
13010250UG 250 μg
130-10-25UG 25 μg
13010500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 130-10-5UG Supplier Gibco™ Supplier No. 130105UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Relaxin-3 is a 5.5 kDa, disulfide-linked, heterodimeric protein consisting of a 24 amino acid A-chain and a 27 amino acid B-chain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Relaxin 3 is part of the Relaxin-like peptide family that plays an important neuropeptide signaling role. Relaxin 3 is a ligand for two structurally related G protein-coupled receptors, human Relaxin receptor 3 and Relaxin receptor 4. The greatest levels of Relaxin 3 expression occur in the brain, specifically in the pons/medulla, with highest levels in the pars ventromedialis of the dorsal tegmental nucleus and with lower levels in the hippocampus and olfactory regions. Significant expression is also observed in the spleen, thymus, lung and ovary. Relaxin 3 inhibits Forskolin-stimulated cAMP accumulation in a dose-dependent manner in cells that express Relaxin receptor 3. Acute and repeated administration of Relaxin 3 causes a decrease in levels of the plasma thyroid stimulating hormone, suggesting that Relaxin 3 may play a role in long-term control of food intake.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8WXF3
For Use With (Application) Control
Formulation Protein with no preservative
Gene ID (Entrez) 117579
Molecular Weight (g/mol) 5.5 kDa
Name Human Relaxin-3
Purification Method Purified
Quantity 5 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias H3; insl7; insulin-like 7; insulin-like peptide 7; Insulin-like peptide INSL7; M3; PRO20213; Prorelaxin H3; prorelaxin M3; relaxin 3; relaxin-3; Relaxin-3 A chain; Relaxin-3 B chain; RLN3; RXN3; UNQ6188; UNQ6188/PRO20213; ZINS4
Common Name Relaxin 3
Gene Symbol RLN3
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence Alpha chain: DVLAGLSSSC CKWGCSKSEI SSLC Beta chain: RAAPYGVRLC GREFIRAVIF TCGGSRW
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.