Learn More
Biomatik Corporation Human Relaxin (RLN) ELISA Kit
Supplier:  Biomatik Corporation EKU07060
Encompass_Preferred
Human Relaxin (RLN) ELISA Kit for detecting RLN in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.2-2,000pg/ml. Sensitivity: 1.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P04808. Gene ID: 6013. Target Synonyms: RLXH1; Relaxin H1; Prorelaxin H1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7323