Learn More
Sino Biological Human RELT ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502372292
Encompass_Preferred
Each vial contains 175 ng of recombinant Human RELT / TNFRSF19L. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2500 pg/mL is recommended.
Catalog No. 50-237-2292 Supplier Sino Biological Supplier No. SEK105305
Each vial contains 175 ng of recombinant Human RELT / TNFRSF19L. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2500 pg/mL is recommended.