Learn More
Sino Biological Recombinant Anti-RELT/TNFRSF19L Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10530R13520
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human RELT / TNFRSF19L (rh RELT / TNFRSF19L; Catalog#10530-H08H; NP_116260.2; Met 1-Ala 160).
Catalog No. 50-240-8491