Learn More
Sino Biological Human RELT MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10530MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human RELT / TNFRSF19L (rh RELT / TNFRSF19L; Catalog#10530-H08H; NP_116260.2; Met1-Ala160). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1229