Learn More
Sino Biological Anti-RELT/TNFRSF19L Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10530RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human RELT (rh RELT; Catalog#10530-H08H; NP_116260.2; Met 1-Ala 160). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0152