Learn More
Sino Biological Anti-RELT/TNFRSF19L Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10530RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human RELT (rh RELT; Catalog#10530-H08H; NP_116260.2; Met 1-Ala 160). RELT specific IgG was purified by human RELT affinity chromatography.
Catalog No. 50-241-0114