Learn More
Sino Biological Anti-Renin Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10969MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Renin / REN (rh Renin / REN; Catalog#10969-H08H; NP_000528.1; Met1-Arg406). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-5014