Learn More
Sino Biological Anti-Renin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10969T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Renin (rh Renin; Catalog#10969-H08H; NP_000528.1; Met1-Arg406). Renin specific IgG was purified by Human Renin affinity chromatography.
Catalog No. 50-255-5011