Learn More
Biomatik Corporation Human Renin (REN) CLIA Kit
Supplier:  Biomatik Corporation EKU08923
Encompass_Preferred
Human Renin (REN) CLIA Kit for detecting REN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 5.49-4,000pg/ml. Sensitivity: 2.24pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P00797. Gene ID: 5972. Target Synonyms: angiotensinogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5937