Learn More
Biomatik Corporation Human Replication Initiator 1 (REPIN1) ELISA Kit
Supplier:  Biomatik Corporation EKU07078
Encompass_Preferred
Human Replication Initiator 1 (REPIN1) ELISA Kit for detecting REPIN1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.129ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BWE0. Gene ID: 29803. Target Synonyms: AP4; RIP60; ZNF464; Zfp464; Replication Initiation Region Protein (60kD) ; Zinc Finger Protein 464; 60 kDa origin-specific DNA-binding protein; ATT-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6934