Learn More
Biomatik Corporation Human Replication Protein A1 (RPA1) ELISA Kit
Supplier:  Biomatik Corporation EKU07080
Encompass_Preferred
Human Replication Protein A1 (RPA1) ELISA Kit for detecting RPA1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27694. Gene ID: 6117. Target Synonyms: HSSB; REPA1; RF-A; RP-A; RPA70; Replication protein A 70 kDa DNA-binding subunit; Single-stranded DNA-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6901