Learn More
Biomatik Corporation Human Resistin (RETN) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12158
Encompass_Preferred
Human Resistin (RETN) Mini sample ELISA Kit for detecting RETN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9HD89. Gene ID: 56729. Target Synonyms: ADSF; FIZZ3; RETN1; RSTN; XCP1; Adipose tissue-specific secretory factor; C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein; Cysteine-rich secreted FIZZ3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4985