Learn More
ATCC Human resp. sync. virus; Strain: Long; RNA
Supplier:  ATCC VR26D
Encompass
This product is a preparation of viral nucleic acids, including viral genomic RNA, in total RNA isolated from Hep-2 cells (ATCC CCL-23) infected with Human respiratory syncytial virus strain Long (ATCC VR-26). Virus and host cells are also available through the ATCC Catalog. This product was prepared using methods known to inactivate viruses. RNA isolated from virally infected cells is appropriate for RT-PCR and other molecular virology applications.
Catalog No. 50-238-1684