Learn More
Sino Biological Human respiratory syncytial virus (RSV) (strain A2) Glycoprotein G Protein (His Tag)
Supplier:  Sino Biological 40830V08H100
Encompass_Preferred
A DNA sequence encoding the human respiratory syncytial virus (RSV) (strain A2) glycoprotein G (P03423) (Ser64-Gln298) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6503