Learn More
Sino Biological Human respiratory syncytial virus (RSV) (B, strain 18537) Glycoprotein G Protein (His Tag)
Supplier:  Sino Biological 40829V08H100
Encompass_Preferred
A DNA sequence encoding the Human respiratory syncytial virus (RSV) (B, strain 18537) Glycoprotein G (P20896) (His67-Asn292) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6502