Learn More
Sino Biological Human respiratory syncytial virus (RSV) Fusion glycoprotein / RSV-F Neutralizing Antibody
Supplier:  Sino Biological 11049R338200
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human respiratory syncytial virus (RSV) Fusion glycoprotein / RSV-F (Catalog#11049-V08B; P03420.1; Met1-Thr529) and was produced using recombinant antibody technology.
Catalog No. 50-224-7380