Learn More
ATCC Human respiratory; syncytial virus; Strain: 9320, RNA
Supplier:  ATCC VR955D
Encompass
Genomic RNA isolated HEp-2 cells (ATCC CCL-23) infected with Human respiratory syncytial virus strain 9320 (ATCC VR-955). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-3289
 Restricted Item