Learn More
ATCC Human respiratory; syncytial virus; Strain: 18537
Catalog No. 502382238
Encompass
Human respiratory syncytial virus strain 18537 is propagated in HEp-2 cells (ATCC CCL-23). This strain was isolated in 1962 from the respiratory secretions of a child with acute respiratory disease seen at Children’s National Hospital in Washington, DC, and was deposited by the National Institute of Allergy and Infectious Diseases (NIAID). It has applications in respiratory disease research.
Catalog No. 50-238-2238 Supplier ATCC Supplier No. VR1580
Human respiratory syncytial virus strain 18537 is propagated in HEp-2 cells (ATCC CCL-23). This strain was isolated in 1962 from the respiratory secretions of a child with acute respiratory disease seen at Children’s National Hospital in Washington, DC, and was deposited by the National Institute of Allergy and Infectious Diseases (NIAID). It has applications in respiratory disease research.