Learn More
ATCC Human respiratory; syncytial virus; Strain: A2 Purified
Catalog No. 502383388
Encompass
Human respiratory syncytial virus, Purified strain A2 is propagated in HeLa cells (ATCC CCL-2). This strain was isolated in 1961 from the lower respiratory tract of an infant with bronchiolitis and bronchopneumonia in Melbourne, Australia. This product is provided as a purified preparation and has applications in respiratory disease research and assay development.
Catalog No. 50-238-3388 Supplier ATCC Supplier No. VR1540P
Human respiratory syncytial virus, Purified strain A2 is propagated in HeLa cells (ATCC CCL-2). This strain was isolated in 1961 from the lower respiratory tract of an infant with bronchiolitis and bronchopneumonia in Melbourne, Australia. This product is provided as a purified preparation and has applications in respiratory disease research and assay development.