Learn More
ATCC Human respiratory; syncytial virus; Strain: B WV/14617/85
Catalog No. 502383849
Encompass
Human respiratory syncytial virus strain B WV/14617/85 is propagated in Vero cells (ATCC CCL-81). This viral strain was isolated in the secretion from the endotracheal tube of an infant with pneumonia and was deposited by the National Institutes of Allergy and Infectious Diseases. It has applications in respiratory disease research.
Catalog No. 50-238-3849 Supplier ATCC Supplier No. VR1400
Human respiratory syncytial virus strain B WV/14617/85 is propagated in Vero cells (ATCC CCL-81). This viral strain was isolated in the secretion from the endotracheal tube of an infant with pneumonia and was deposited by the National Institutes of Allergy and Infectious Diseases. It has applications in respiratory disease research.