Learn More
ATCC Human respiratory; syncytial virus RNA; Strain: ATCC-2012-11
Supplier: ATCC VR1803D
Genomic RNA isolated from a preparation of cell lysate and supernatant from Hep-2 cells (ATCC CCL-23) infected with Human respiratory syncytial virus strain ATCC-2012-11 (ATCC VR-1803). This product was prepared using methods known to inactivate viruses. The source organism and host cells are also available through the ATCC catalog.