Learn More
Sino Biological Human respiratory syncytial virus (RSV) (A2) Fusion glycoprotein F0 Peptide Pool 1 vial (50 μg/peptide)
Supplier:  Sino Biological PP0041
Encompass_Preferred
The Human respiratory syncytial virus (RSV) (A2) Fusion glycoprotein F0 Peptide Pool, consisting mainly of 15-mer sequences with 11 amino acids (aa) overlap.
Catalog No. 50-006-96187